Description
- Two-part, translucent silicone system used with injection molding equipment
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a low durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
- Suitable for over-molding applications
- Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
Product Details
Property | Average Result |
---|---|
Durometer | 20 Type A |
Tensile | 750 psi (5.17 MPa) |
Elongation | 700 % |
Tear | 125 ppi (22.05 kN/m) |
Appearance | Translucent |
Cure | 5 minutes / 150 °C |
Cure Rate Scorch | 1.5 minutes |
Cure Rate T90 | 2.1 minutes |
Cure System | Platinum |
Extrusion Rate | 80 g/minute |
Mix Ratio | 1:1 |
Specific Gravity | 1.14 |
Stress at Strain | 65 psi / 200 % (0.45 MPa / 200 %) |
Work Time | |
Comment | Injection Molding Elastomer |