Implant Line
MED-4860
LIQUID SILICONE RUBBER
Description
  • Two-part, translucent silicone system designed for use with injection molding equipment
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For the injection molding of parts requiring a material with a high durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
  • Suitable for over-molding applications
  • Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
Product Details
Property Average Result
Durometer 60 Type A
Tensile 1,350 psi (9.31 MPa)
Elongation 530 %
Tear 255 ppi (44.97 kN/m)
Appearance Translucent
Cure 5 minutes / 165 °C
Cure System Platinum
Extrusion Rate 85 g/minute
Mix Ratio 1:1
Specific Gravity 1.15
Stress at Strain 600 psi / 200 % (4.14 MPa / 200 %)
Work Time 72 hours
Comment Injection Molding Elastomer