Implant Line
MED-4830
LIQUID SILICONE RUBBER
Description
  • Two-part, translucent silicone system designed for use with injection molding equipment
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For the injection molding of parts requiring a material with a medium durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
  • Suitable for over-molding applications
  • Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
Product Details
Property Average Result
Durometer 30 Type A
Tensile 1,350 psi (9.31 MPa)
Elongation 750 %
Tear 150 ppi (26.46 kN/m)
Appearance Translucent
Cure 5 minutes / 150 °C
Cure System Platinum
Extrusion Rate 175 g/minute
Mix Ratio 1:1
Specific Gravity 1.13
Stress at Strain 175 psi / 200 % (1.21 MPa / 200 %)
Work Time
Comment Injection Molding Elastomer