Description
- Two-part, high consistency silicone elastomer
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
- For applications requiring high physical properties and fatigue and abrasion resistance
Product Details
Property | Average Result |
---|---|
Durometer | 70 Type A |
Tensile | 1,415 psi (9.76 MPa) |
Elongation | 760 % |
Tear | 300 ppi (52.91 kN/m) |
Appearance | Translucent |
Cure | 10 minutes / 138 °C |
Cure System | Platinum |
Mix Ratio | 1:1 |
Plasticity | |
Post Cure | 4 hours / 177 °C |
Special Feature | |
Specific Gravity | 1.19 |
Stress at Strain | 595 psi / 200 % (4.1 MPa / 200 %) |
Work Time | 2.5 hours |
Comment | Abrasion / Fatigue Resistant |