Description
- Two-part, high tear strength silicone elastomer
- Soft silicone
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 35 Type A |
Tensile | 1,535 psi (10.59 MPa) |
Elongation | 1055 % |
Tear | 195 ppi (34.39 kN/m) |
Appearance | Translucent |
Cure | 10 minutes / 116 °C |
Cure System | Platinum |
Mix Ratio | 1:1 |
Plasticity | |
Specific Gravity | 1.11 |
Stress at Strain | 180 psi / 200 % (1.24 MPa / 200 %) |
Work Time | 2 hours |
Comment | High Tear |