Description
- Two-part, low tension set silicone elastomer
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 30 Type A |
Tensile | 1,395 psi (9.62 MPa) |
Elongation | 890 % |
Tear | 135 ppi (23.81 kN/m) |
Appearance | Translucent |
Cure | 10 minutes / 171 °C |
Cure System | Platinum |
Mix Ratio | 1:1 |
Plasticity | |
Specific Gravity | 1.11 |
Stress at Strain | 110 psi / 200 % (0.76 MPa / 200 %) |
Work Time | 2 hours |
Comment | Low Tension Set |