Description
- Two-part, high tear strength silicone elastomer
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 15 Type A |
Tensile | 715 psi (4.93 MPa) |
Elongation | 1385 % |
Tear | 155 ppi (27.34 kN/m) |
Appearance | Translucent |
Cure | 10 minutes / 116 °C |
Cure System | Platinum |
Mix Ratio | 1:1 |
Plasticity | |
Specific Gravity | 1.08 |
Stress at Strain | 40 psi / 200 % (0.28 MPa / 200 %) |
Work Time | 26 hours |
Comment | Low Durometer |