Description
- Uncatalyzed, vinyl-functional, high consistency silicone elastomer
- Medium durometer
- Requires the addition of a peroxide catalyst to accomplish vulcanization
- Strained through a 400-mesh screen (minimum)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 50 Type A |
Tensile | 1,400 psi (9.66 MPa) |
Elongation | 750 % |
Tear | 195 ppi (34.39 kN/m) |
Appearance | Translucent |
Cure | 5 minutes / 116 °C |
Cure System | Peroxide |
Mix Ratio | 1 PART |
Plasticity | |
Post Cure | 2 hours / 177 °C |
Specific Gravity | 1.16 |
Stress at Strain | 250 psi / 200 % (1.72 MPa / 200 %) |
Comment | Uncatalyzed |