Implant Line
MED-4502-6
COLOR MASTERBATCH FOR HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • A green, single component masterbatch, with the consistency of a clay like material
  • Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
  • Supported by USP Class VI and ISO 10993 Biological testing (reference Biological Testing Data Table)
Applications
  • For easy and more precise pigmentation of high consistency silicone materials, suitable for molding, calendering or extrusion
  • Reduces production time and eliminates use of powders that may contaminate clean room environments
  • Strict quality controls ensure color consistency
Product Details
Property Average Result
Appearance Green
Mix Ratio 1 PART
Comment Masterbatch to add color to HCR platinum cure systems