MED-4502-1
COLOR MASTERBATCH FOR HIGH CONSISTENCY SILICONE ELASTOMERS
Description
- A white, single component masterbatch, with the consistency of a low viscosity paste a clay like material
- Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
- Supported by USP Class VI and ISO 10993 Biological testing (reference Biological Testing Data Table)
Applications
- For easy and more precise pigmentation of high consistency silicone materials, suitable for molding, calendering or extrusion
- Reduces production time and eliminates use of powders that may contaminate clean room environments
- Strict quality controls ensure color consistency
Product Details
Property | Average Result |
---|---|
Appearance | White |
Mix Ratio | 1 PART |
Comment | Masterbatch to add color to HCR platinum cure systems |