Implant Line
MED-4286
ULTRA-SOFT LOW VISCOSITY SILICONE ELASTOMER
Description
  • Two-part, translucent silicone elastomer
  • Cures via addition-cure chemistry
  • Requires heat to cure
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For potting, encapsulating or coating electronics and other components
  • For use with transfer or low-pressure injection molding processes
NuSil® MED-4286 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Viscosity 6,350 cP (6,350 mPa*s)
Durometer 55 Type 000
Work Time 17.5 hours
Tensile 45 psi (0.31 MPa)
Cure 45 minutes / 150 °C
Cure System Platinum
Cure Type Heat Cure
Elongation 475 %
Mix Ratio 1:1
Stress at Strain 8 psi / 200 % (0.06 MPa / 200 %)