Implant Line
MED-4244
LOW CONSISTENCY SILICONE ELASTOMER
Description
  • Two-part, translucent, pourable silicone elastomer
  • Cures with heat via addition-cure chemistry
  • 10:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
NuSil Technology’s MED-4244 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 40 Type A
Viscosity 130,000 cP (130,000 mPa*s)
Work Time 4 hours
Tensile 865 psi (5.97 MPa)
Appearance Translucent
Cure 3 minutes / 177 °C
Cure System Platinum
Elongation 350 %
Mix Ratio 10:1
Post Cure 1 hour / 150 °C
Specific Gravity 1.1
Stress at Strain 480 psi / 200 % (3.31 MPa / 200 %)
Tear 150 ppi (26.46 kN/m)
Comment Flowable, Medium Durometer