Description
- A two-part, translucent pourable silicone elastomer
- Cures with heat via addition-cure chemistry
- 10:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | 25 Type A |
Viscosity | 97,800 cP (97,800 mPa*s) |
Tensile | 685 psi (4.72 MPa) |
Appearance | Translucent |
Cure | 3 minutes / 150 °C |
Cure System | Platinum |
Elongation | 530 % |
Mix Ratio | 10:1 |
Post Cure | 1 hour / 150 °C |
Specific Gravity | 1.1 |
Comment | Flowable |