Description
- An uncatalyzed high consistency material
- Catalyze with various peroxides to form a medium durometer rubber
- Strained through fine mesh stainless steel screens to ensure freedom from particulate contamination
Applications
- For fabricating molded and extruded silicone rubber parts
Product Details
Property | Average Result |
---|---|
Durometer | 50 Type A |
Tensile | 1,200 psi (8.28 MPa) |
Elongation | 775 % |
Tear | 225 ppi (39.68 kN/m) |
Appearance | Translucent |
Cure | 5 minutes / 116 °C |
Cure Rate Scorch | 0.55 minutes |
Cure Rate T90 | 1.7 minutes |
Cure System | Peroxide |
Mix Ratio | 1 PART |
Plasticity | |
Post Cure | 4 hours / 205 °C |
Specific Gravity | 1.15 |
Stress at Strain | 325 psi / 200 % (2.24 MPa / 200 %) |
Comment | Uncatalyzed |