Description
- One-part, silicone dispersion containing high molecular weight hydroxyl-functional silicone polymer dispersed in xylene Non-curing
Applications
- Suitable for dip casting of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
- Ideal for use as a needle coating
- Provides excellent lubricity for cutting edges, needle cannula, and other applications requiring friction reduction
Product Details
Property | Average Result |
---|---|
Cure System | Non-Curing |
Viscosity | 27,000 cP (27,000 mPa*s) |
Non Volatile Content | 31 % |
Appearance | Translucent |
Mix Ratio | 1 PART |
Solvent | Xylene |
Special Feature | |
Comment | Lubricious Coating |