Description
- Uncatalyzed, vinyl-functional, high consistency silicone elastomer
- Requires the addition of a peroxide catalyst to accomplish vulcanization
- Low tack surface
- Strained through a 400-mesh screen (minimum)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding of parts
- Ideal for applications requiring a material with a low durometer, including: gaskets, seals, molded balloons, and catheter tips
Product Details
Property | Average Result |
---|---|
Durometer | 25 Type A |
Tensile | 1,010 psi (6.97 MPa) |
Elongation | 800 % |
Tear | 70 ppi (12.35 kN/m) |
Appearance | Translucent |
Cure | 10 minutes / 116 °C |
Cure Rate Scorch | 0.55 minutes |
Cure Rate T90 | 1.65 minutes |
Cure System | Peroxide |
Mix Ratio | 1 PART |
Plasticity | |
Post Cure | 2 hours / 200 °C |
Specific Gravity | 1.11 |
Stress at Strain | 90 psi / 200 % (0.62 MPa / 200 %) |
Comment | Low Tension Set, Uncatalyzed |