Description
- Uncatalyzed, vinyl-functional, high consistency silicone elastomer
- Requires addition of a peroxide catalyst for vulcanization
- Strained through a 400-mesh screen (minimum) to ensure freedom from particulate contamination
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 30 Type A |
Tensile | 1,300 psi (8.97 MPa) |
Elongation | 950 % |
Tear | 140 ppi (24.69 kN/m) |
Appearance | Translucent |
Cure | 5 minutes / 116 °C |
Cure Rate Scorch | 0.55 minutes |
Cure Rate T90 | 2.1 minutes |
Cure System | Peroxide |
Mix Ratio | 1 PART |
Plasticity | |
Post Cure | 2 hours / 177 °C |
Specific Gravity | 1.1 |
Stress at Strain | 100 psi / 200 % (0.69 MPa / 200 %) |
Comment | Uncatalyzed |