Description
- Two-part, high durometer, tear resistant silicone elastomer
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
- For application uses requiring high durometer and high-modulus characteristics
Product Details
Property | Average Result |
---|---|
Durometer | 80 Type A |
Tensile | 1,150 psi (7.93 MPa) |
Elongation | 700 % |
Tear | 225 ppi (39.68 kN/m) |
Appearance | Translucent Tan |
Cure | 10 minutes / 116 °C |
Cure Rate Scorch | 0.8 minutes |
Cure Rate T90 | 2.75 minutes |
Cure System | Platinum |
Mix Ratio | 1:1 |
Plasticity | |
Specific Gravity | 1.2 |
Stress at Strain | 450 psi / 200 % (3.1 MPa / 200 %) |
Work Time | 8 hours |
Comment | High Tear Strength |