Description
- Two-part, high tear strength silicone elastomer
- Firm silicone
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 65 Type A |
Tensile | 1,250 psi (8.62 MPa) |
Elongation | 1000 % |
Tear | 250 ppi (44.09 kN/m) |
Appearance | Translucent Tan |
Cure | 10 minutes / 116 °C |
Cure Rate Scorch | 0.85 minutes |
Cure Rate T90 | 2.65 minutes |
Cure System | Platinum |
Mix Ratio | 1:1 |
Plasticity | |
Specific Gravity | 1.2 |
Stress at Strain | 350 psi / 200 % (2.41 MPa / 200 %) |
Work Time | 6 hours |
Comment | High Tear Strength |