Description
- Two-part, high tear strength silicone elastomer
- Medium durometer silicone
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 50 Type A |
Tensile | 1,500 psi (10.34 MPa) |
Elongation | 985 % |
Tear | 250 ppi (44.09 kN/m) |
Appearance | Translucent Gray |
Cure | 10 minutes / 115 °C |
Cure Rate Scorch | 1.1 minutes |
Cure Rate T90 | 2.6 minutes |
Cure System | Platinum |
Mix Ratio | 1:1 |
Plasticity | |
Specific Gravity | 1.16 |
Stress at Strain | 300 psi / 200 % (2.07 Mpa / 200 %) |
Work Time | 3.5 hours |
Comment | High Tear Strength |