Description
- A unique three-part system, VersaSil3 offers the flexibility of adjustable cure rate and table life for various fabrication requirements
- High tear strength, wide processing parameters, and translucent, non-tacky surfaces
- Developed to be compounded with the inhibitor CAT-40 and the platinum catalyst CAT-55. Advantages include: no volatile by-products, lower cure temperature, and optional post-cure
- 100: 1.0: 1.0 Mix Ratio
Applications
- For mass production with maximum flexibility
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 40 Type A |
Tensile | 1,500 psi (10.34 MPa) |
Elongation | 1000 % |
Tear | 160 ppi (28.22 kN/m) |
Cure | 10 minutes / 116 °C |
Cure Rate Scorch | 0.8 minutes |
Cure Rate T90 | 2.45 minutes |
Mix Ratio | 100:1:1 |
Special Feature | |
Specific Gravity | 1.11 |
Stress at Strain | 230 psi / 200 % (1.59 MPa / 200 %) |
Work Time | 2 hours |
Comment | Can be Cured via Platinum or Peroxide |