Description
- High tensile strength, low modulus high consistency silicone rubber
- Low tensile set and low hysteresis qualities
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
- Ideal for applications requiring low modulus and high strength properties such as balloons and bladders
- Can be considered as a replacement material for latex/natural rubber
Product Details
Property | Average Result |
---|---|
Durometer | 30 Type A |
Tensile | 2,300 psi (15.86 MPa) |
Elongation | 1,050 % |
Tear | 235 ppi (41.45 kN/m) |
Appearance | Translucent |
Cure | 10 minutes / 171 °C |
Mix Ratio | 1:1 |
Specific Gravity | 1.11 |
Stress at Strain | 100 psi / 200 % (0.69 MPa / 200 %) |
Work Time | 2.5 hours |
Comment | Ultra-high tensile strength |