Description
- Two-part, translucent gray, pourable silicone system
- Cures with heat via addition-cure chemistry
- 10:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | 27 Type A |
Viscosity | 100,000 cP |
Tensile | 670 psi (4.62 MPa) |
Appearance | Translucent |
Cure | 3 minutes / 150 °C |
Cure System | Platinum |
Elongation | 530 % |
Mix Ratio | 10:1 |
Post Cure | 1 hours / 150 °C |
Specific Gravity | 1.09 |
Comment | Flowable |