Description
- A clear liquid trifluoroprophylmethylsiloxane to provide lubricious and/or hydrophobic coating, for lubricating the surfaces of molded dimethyl silicone elastomer parts
- Available in standard viscosities of : 350 cP, 1,000 cP, and 12,500 cP.
- Custom viscosities available upon request
Applications
- For applications requiring a material that is highly water repellent and resists decomposition from heat and oxidation
- Has reduced solubility, good wetting properties and good lubricative characteristics in contact with plastics, rubber and skin
Product Details
Property | Average Result |
---|---|
Volatile Content | 0.3 % |
Viscosity | 350 cP / 1,000 cP / 12,500 cP (350 mPa*s / 1,000 mPa*s / 12,500 mPa*s) |
Substrate | Silicone |
Comment | Fluorosilicone Polymer |