Description
- A transparent, colorless polydimethylsiloxane liquid
- Available in standard viscosities of: 1,000 cP
- Custom viscosities available upon request
Applications
- For applications requiring a material that is highly water repellent and resists decomposition from heat and oxidation
- Has good wetting properties and good lubricative characteristics in contact with plastics, rubber and skin
Product Details
Property | Average Result |
---|---|
Volatile Content | 0.1 % |
Viscosity | 1,000 cP (1,000 mPa*s) |
Appearance | Clear |
Cure System | Non-Curing |
Mix Ratio | 1 Part |
Refractive Index | 1.40 |
Specific Gravity | 0.97 |
Comment | Highly purified Dimethyl Polymer |