Description
- A clear polydimethylsiloxane liquid to provide a lubricious and/or hydrophobic coating
- Available in custom viscosities of 100 – 100,000 cP
- Has controlled volatility, lubrication characteristics and low surface tension
- Standard viscosities of 100, 350, 1000, and 12,500 cP
Applications
- For applications requiring a material that is highly water repellent and resists decomposition from heat and oxidation
- Has good wetting properties and good lubricative characteristics in contact with plastics, rubber and skin
Note: MED-361 is not available for any long-term ophthalmic application
Product Details
Property | Average Result |
---|---|
Volatile Content | 0.1 % |
Viscosity | 100 cP / 350 cP / 1,000 cP / 12,500 cP (100 mPa*s / 350 mPa*s / 1,000 mPa*s / 12,500 mPa*s) |
Appearance | Clear |
Substrate | Glass / Metal / Plastic |
Comment | Dimethyl Polymer |