Description
- Two-part, room temperature curing, silicone foam
- Platinum cure
- No odor
- Yields a low density silicone foam when catalyzed
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Can be used to make foam sheeting or ribbon
- Use in shock and vibration dampening situations requiring a lightweight, flexible foam
- Device applications requiring a low density component
Product Details
Property | Average Result |
---|---|
Viscosity | 40,000 cP (40,000 mPa*s) |
Cure System | Platinum |
Application Life | 2 minutes |
Appearance | White |
Mix Ratio | 1:1 |
Comment | Low Density |