Implant Line
MED-2214
SILICONE DISPERSION
Description
  • One-part, platinum-catalyzed, siloxane elastomer dispersed in xylene
  • Heat-cures to form strong, thin, elastomeric films
  • Excellent physical properties
  • Usable life can be extended with refrigeration
Applications
  • Suitable for dip casting and heat-curing of thin elastomeric films
  • For use as an adhesive for bonding and sealing silicone together
  • Low viscosity makes dispersions ideal for use as sprayable coatings
NuSil Technology’s MED-2214 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Cure System Platinum
Viscosity 3,700 cP (3,700 mPa*s)
Non Volatile Content 35 %
Durometer 35 Type A
Appearance Translucent
Cure 60 minutes / 50 °C
Elongation 850 %
Mix Ratio 1 Part
Tear 170 ppi (29.98 kN/m)
Tensile 1,700 psi (11.72 MPa)
Comment One Part, Requires heat to cure