Description
- Uncatalyzed high consistency elastomer
- Catalyze with various peroxides to form a medium durometer rubber
- Strained through a 400-mesh screen (minimum)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
Product Details
Property | Average Result |
---|---|
Durometer | 50 Type A |
Tensile | 1,300 psi (8.97 MPa) |
Elongation | 750 % |
Tear | 225 ppi (39.68 kN/m) |
Appearance | Clear |
Cure | 5 minutes / 116 °C |
Cure System | Peroxide |
Mix Ratio | 1 PART |
Plasticity | |
Post Cure | 4 hours / 205 °C |
Specific Gravity | 1.15 |
Stress at Strain | 325 psi / 200 % (2.24 MPa / 200 %) |
Comment | Uncatalyzed |