Description
- Fully compounded, platinum-catalyzed, dimethyl-methylvinyl siloxane elastomers dispersed in xylene
- Excellent physical properties
- Usable life can be extended with refrigeration
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- For use as an adhesive for bonding and sealing silicone together
- Low viscosity makes dispersions ideal for use as sprayable coatings
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 1,800 cP (1,800 mPa*s) |
Non Volatile Content | 35 % |
Appearance | Translucent |
Cure | 60 minutes / 50 °C 60 minutes / 150 °C 3 minutes / RTV |
Elongation | 900 % |
Mix Ratio | 1 PART |
Solvent | Xylene |
Specific Gravity | |
Stress At Strain | |
Tear | 160 ppi (28.22 kN/m) |
Tensile | 1,700 psi (11.72 MPa) |
Comment | One Part, Requires heat to cure |