Premium Care Line
MED-165
SILICONE PRIMER
Description
  • Easy to use, air-drying, one-part primer designed for use with platinum-cured systems where conventional silicone primers are insufficient
  • One-component primer supplied in VM&P Naphtha requires no mixing
  • Flash Point (TCC) of 17°C (63°F)
  • Convenient container sizes yield less waste
Applications
  • Improves adhesion of silicones to various substrates including: metals (such as stainless steel, steel, copper and aluminum), ceramics, rigid plastics, and other silicone materials
NuSil Technology’s MED-165 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Cure 1 hours / RTV
Mix Ratio 1 PART
Non Volatile Content 4.8 %
Specific Gravity 0.77
Appearance Translucent
Lap Shear 515 psi (3.55 MPa)
Solvent Naptha
Comment Increased adhesion to various metals.