Description
- Specially formulated primer
- One-component primer supplied in VM&P Naphtha requires no mixing
- Easy to use, air-drying, one-part primer designed for use with one and two-part RTV systems where conventional silicone primers are insufficient
- Flash Point (TCC) of 17°C (63°F)
- Convenient container sizes yield less waste
Applications
- Improves adhesion of silicones to various substrates including: metals (such as stainless steel, steel, copper and aluminum), ceramics, rigid plastics, and other silicone materials
Product Details
Property | Average Result |
---|---|
Cure | 1 hours / RTV |
Mix Ratio | 1 PART |
Non Volatile Content | 4 % |
Specific Gravity | 0.77 |
Appearance | Translucent |
Lap Shear | 330 psi (2.28 MPa) |
Solvent | Naptha |
Comment | General all-purpose primer. Use with platinum or moisture cure silicones. |