Description
- One-part, translucent, self-leveling silicone adhesive
- Cures at room temperature upon exposure to atmospheric moisture
- Supplied ready to use in Semco® cartridges
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
Product Details
Property | Average Result |
---|---|
Cure Type | RTV |
Mix Ratio | 1 PART |
Rheology | Flowable / Self Leveling |
Appearance | Translucent |
Cure | 72 hours / RTV |
Cure System | Acetoxy |
Durometer | 23 Type A |
Elongation | 340 % |
Flow Rate | 2.7 in (68.58 mm) |
Specific Gravity | 1.03 |
Substrate | Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium |
Tack Free Time | 9.5 minutes |
Tear | 17 ppi (3 kN/m) |
Tensile | 265 psi (1.83 MPa) |
Comment | High flow |