Description
- One-part, solvent-free silicone
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
Product Details
Property | Average Result |
---|---|
Cure Type | RTV |
Mix Ratio | 1 PART |
Rheology | Flowable / Self Leveling |
Appearance | Translucent |
Cure | 72 hours / RTV |
Cure System | Acetoxy |
Durometer | 25 Type A |
Elongation | 680 % |
Extrusion Rate | 100 g/minute |
Specific Gravity | 1.12 |
Stress at Strain | 130 psi / 200 % (0.9 MPa / 200 %) |
Substrate | Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate / Stainless steel |
Tack Free Time | 12 minutes |
Tear | 95 ppi (16.75 kN/m) |
Tensile | 1,440 psi (9.93 MPa) |
Comment | Low flow |