Description
- One-part, non-curing, pressure sensitive adhesive
- Supplied in ethyl acetate
Applications
- For temporarily adhering various substrates (including metals, glass, silicone, organic rubbers, and skin)
- Use for medical tapes and fasteners, external incontinence devices, transdermal skin patches, hair replacement adhesives, and special effects adhesives
- Non-irritating to skin
Product Details
Property | Average Result |
---|---|
Viscosity | 3,500 cP (3,500 mPa*s) |
Appearance | Translucent |
Cure | 60 minutes / 85 °C |
Volatile Content | |
Comment | High cohesive strength |