Implant Line
MED-1131
RTV SILICONE ADHESIVE
Description
  • One part, low slump, solvent-free translucent silicone
  • Cures at room temperature upon exposure to atmospheric moisture
Applications
  • Well suited for use as an adhesive for bonding and sealing silicone materials
  • For applications requiring the bonding of silicone to metals, urethanes and various other substrates
NuSil Technology’s MED-1131 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Cure Type RTV
Mix Ratio 1 PART
Rheology Thixotropic / non-slump
Appearance Translucent
Cure 7 days / RTV
Cure System Oxime
Durometer 35 Type A
Elongation 330 %
Flow Rate 0.4 in (10.16 mm)
Specific Gravity 1.1
Substrate Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium
Tack Free Time 20 minutes
Tear 45 ppi (7.94 kN/m)
Tensile 890 psi (6.14 MPa)
Comment Non-acidic leaving group