Description
- One part, low slump, solvent-free translucent silicone
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
Product Details
Property | Average Result |
---|---|
Cure Type | RTV |
Mix Ratio | 1 PART |
Rheology | Thixotropic / non-slump |
Appearance | Translucent |
Cure | 72 hours / RTV |
Cure System | Acetoxy |
Durometer | 30 Type A |
Elongation | 565 % |
Extrusion Rate | 2.8 g/minute |
Flow Rate | 0.2 in (5.08 mm) |
Specific Gravity | 1.07 |
Stress at Strain | 170 psi / 200 % (1.17 MPa / 200 %) |
Substrate | Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate |
Tack Free Time | 9 minutes |
Tensile | 730 psi (5.03 MPa) |
Comment | Low-slump, solvent-free translucent silicone |