Description
- One part, solvent-free, non-slumping translucent silicone
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
Product Details
Property | Average Result |
---|---|
Cure Type | RTV |
Mix Ratio | 1 PART |
Rheology | Thixotropic / non-slump |
Appearance | Translucent |
Cure | 7 days / RTV |
Cure System | Oxime |
Durometer | 35 Type A |
Elongation | 310 % |
Extrusion Rate | 70 g/minute |
Specific Gravity | 1.1 |
Stress at Strain | 415 psi / 200 % (2.86 MPa / 200 %) |
Substrate | Primerless: Glass / Polyurethane / Aluminum / Silicone / Titanium |
Tack Free Time | 22 minutes |
Tear | 40 ppi (7.05 kN/m) |
Tensile | 840 psi (5.79 MPa) |
Comment | Non-acidic leaving group |