Description
- One part, solvent-free silicone
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Well suited for use as an adhesive for bonding and sealing silicone materials
- For applications requiring the bonding of silicone to metals, urethanes and various other substrates
Product Details
Property | Average Result |
---|---|
Cure Type | RTV |
Mix Ratio | 1 PART |
Rheology | Flowable / Self Leveling |
Appearance | Translucent |
Cure | 72 hours / RTV |
Cure System | Acetoxy |
Durometer | 25 Type A |
Elongation | 795 % |
Extrusion Rate | 2.5 g/minute |
Specific Gravity | 1.08 |
Stress At Strain | |
Substrate | Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate |
Tack Free Time | 12 minutes |
Tear | 75 ppi (13.23 kN/m) |
Tensile | 1,400 psi (9.66 MPa) |
Comment | Medium flow, high extrusion rate |