Description
- Two-part, optically clear silicone system
- Based on a methyl silicone polymer
- Low viscosity
- 1:1 Mix Ratio (Part A:B)
Applications
- For bonding, casting or injection molding of high performance optical components
- For applications requiring index matching at 1.40
- For photonics applications requiring low outgassing and minimal volatile condensables to avoid condensation in sensitive devices
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 3,125 cP (3,125 mPa*s) |
Durometer | 50 Type A |
Tensile | 850 psi (5.86 MPa) |
Elongation | 90 % |
Appearance | Clear |
Cte | |
Cure | 15 minutes / 150 °C |
Dielectric Strength | 520 volts/mil (20.28 kV/mm) |
Ionic Content Cl | <5 ppm |
Ionic Content K | <2 ppm |
Ionic Content Na | <4 ppm |
Lap Shear | 200 psi (1.38 MPa) |
Mix Ratio | 1:1 |
Refractive Index | 1.40 |
Specific Gravity | 1.02 |
Tack Free Time | 6 hours |
Volume Resistivity | 1,100,000,000,000,000 ohmcm |
Work Time | 3 hours |
Comment | RTV; Low volatility |