Description
- High purity, optically clear, medium to firm silicone gel
- Two components blend easily in a convenient 1:1 ratio by weight or volume
Applications
- For potting, encapsulating, backfilling, and dampening applications requiring a medium to firm gel with optical clarity
- For photonics applications including HBLED’s, photo detectors, flat panel displays and tacking/staking sensitive joint
- For applications requiring an operating temperature range of -65°C to 200°C
- (-85°F to 392°F)
Product Details
Property | Average Result |
---|---|
Viscosity | 600 cP (600 mPa*s) |
Penetration | 9 mm |
Appearance | Translucent |
Cure | 90 minutes / 80 °C |
Cure System | Platinum |
Dielectric Strength | 500 volts/mil (19.5 kV/mm) |
Mix Ratio | 1:1 |
Operating Temperature | -65 °C / 200 °C |
Volume Resistivity | 1,000,000,000,000,000 ohmcm |
Comment | Very Soft High Tack Gel;Max viscosity after 6 hrs is 2,000 cP |