Description
- High purity, optically clear, soft silicone gel1:1 Mix Ratio (A:B)
Applications
- For potting, encapsulating, backfilling, and dampening applications requiring a soft gel with optical clarity
Product Details
Property | Average Result |
---|---|
Viscosity | 535 cP (535 mPa*s) |
Appearance | Clear |
Cure | 1 hours / 100 °C |
Mix Ratio | 1:1 |
Specific Gravity | 0.98 |