Description
- A stiff methyl silicone grease
- Typical vapor pressure of 10-8 torr
Applications
- For extreme high vacuum and pressure systems
- Specific applications include: glass and ceramic stopcocks, ground glass joints, high temperature gaskets, valves, seals and O-rings
Product Details
Property | Average Result |
---|---|
Viscosity | 980,100 cP (980,100 mPa*s) |
Specific Gravity | 1.11 |
Volatile Content | 0.3 % |
Appearance | Gray |
Extrusion Rate | 53 g/minute |
Operating Temperature | -40 °C / 230 °C |
Comment | A stiff methyl grease used lubricating . 0.3% Weight loss. For high vacuum and pressure systems. |