Description
- A stiff methyl silicone/Teflon? grease
- Stable across a broad temperature range
- Typical vapor pressure of 10-8 torr
- Resistant to chemicals
Applications
- Provides more tack and excellent lubricating and sealing ability under extreme operating conditions
- For extreme high vacuum and pressure systems
- Specific applications include: glass and ceramic stopcocks, ground glass joints, and high temperature gaskets, valves, seals and O-rings
Product Details
Property | Average Result |
---|---|
Viscosity | 1,100,000 cP (1,100,000 mPa*s) |
Specific Gravity | 1.14 |
Volatile Content | 0.34 % |
Appearance | Gray |
Flow Rate | 2.7 in (68.58 mm) |
Operating Temperature | -40 °C / 230 °C |
Comment | A stiff methyl silicone/Teflon grease for high vacuum and pressure systems. |