FS-3730-2
FLUOROSILICONE ADHESIVE/SEALANT
Description
- A black, one-part, RTV silicone
- Based on a fluorosilicone polymer
- 100 mol % fluorosilicone
Applications
- For sealing and bonding applications requiring solvent and/or fuel resistance
- Bonds aggressively to most surfaces
- For protecting or bonding hardware components exposed to gasoline or aviation fuels
Product Details
Property | Average Result |
---|---|
Cure System | Acetoxy |
Extrusion Rate | 200 g/minute |
Lap Shear | 285 psi (1.97 MPa) |
Durometer | 40 Type A |
Appearance | Black |
Attribute | |
Cure | 72 hours / RTV |
Elongation | 315 % |
Mix Ratio | 1 PART |
Non Volatile Content | 100 % |
Specific Gravity | 1.41 |
Tack Free Time | 10 minutes |
Tear | 50 ppi (8.82 kN/m) |
Tensile | 650 psi (4.48 MPa) |
Comment | Lap Shear after 7 days *3) 300 psi (2.1 MPa) |