Description
- Two-part, clear silicone system
- Offers low modulus and low viscosity
- 1:1 Mix Ratio (Part A: Part B)
- RoHS compliant
Applications
- For applications requiring low volatility and high purity
- Designed for protection of electrical components and assemblies against shock, vibration, moisture, dust, chemicals, and other environmental hazards
- Ideal for applications in which clarity and low viscosity are important
Product Details
Property | Average Result |
---|---|
Viscosity | 3,225 cP (3,225 mPa*s) |
Durometer | 50 Type A |
Tensile | 850 psi (5.86 MPa) |
Elongation | 90 % |
Appearance | Clear |
Dielectric Strength | 610 volts/mil (23.79 kV/mm) |
Lap Shear | 250 psi (1.72 MPa) |
Specific Gravity | 1.02 |
Tack Free Time | 5 hours |
Volatile Content | 0.70 % |
Work Time | 3 hours |