CareSil
CXG-1104
ELASTOMER GEL
Description
  • Proprietary process creates an elastomer gel free of pilling and balling
  • High clarity silicone
  • Enhances soft focus attributes and increased cushion
  • Delivers a silky and enhanced powdery feel without cyclics
  • Ultra-mattifying
  • Non-tacky film former that provides excellent coverage
  • Texturizing balance – adds structure while not compromising spreadability
  • Ideal for non-irritating and sensitive skin care application
Applications
  • Moisturization
  • Anti-Aging/Anti-Wrinkle
  • Sunscreen, After- Sun, and Self- Tanners
  • Skin Repair Serums
  • Skin Tone Repair
  • Men’s Grooming
  • Color Cosmetics
Product Details
Property Average Result
INCI Name Dimethicone (and) Dimethicone/Vinyl Dimethicone Crosspolymer
D4 Content <0.05 %
Viscosity 635,000 cP (635,000 mPa*s)
Appearance Clear
Refractive Index 1.40
Specific Gravity 0.92
Comment 12% Solids Elastomer Gel Dispersed in Low Viscosity Dimethicone Fluid