Description
- Electrically conductive, gray silicone grease
- Excellent resistance to moisture, ozone and oxidation over a wide temperature range
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- For electric conductivity, high and low temperature stability, and low bleed
- For providing lubrication and static dissipation
Product Details
Property | Average Result |
---|---|
Specific Gravity | 1.1 |
Appearance | Gray |
Volume Resistivity | 1,000,000,000,000,000 ohmcm |
Comment | Volume Resistivity 1x1015 ohmcm, Thermally conductive |