Description
- Thermally conductive, white silicone grease
- Excellent resistance to moisture, ozone and oxidation over a wide temperature range
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- For thermal conductivity, high and low temperature stability, and low bleed
- Apply to the base and mounting studs of transistors and other electrical components as a heat-sink seal
Product Details
Property | Average Result |
---|---|
Specific Gravity | 1.61 |
Appearance | White |
Comment | Thermally Conductive |