Description
- Two-part, controlled volatility, clear silicone gel
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Useful for potting intricate assemblies due to a low viscosity
- As an embedding or potting compound for environmental protection of electronic assemblies and components in electronic or space applications
Product Details
Property | Average Result |
---|---|
Viscosity | 2,500 cP (2,500 mPa*s) |
Penetration | 4 mm |
Work Time | 30 hours |
Appearance | Clear |
Cure | 30 minutes / 150 °C |
Cure System | Platinum |
Mix Ratio | 1:1 |
Comment | Low Viscosity, Penetration 4.0 mm |